Learn More
TRI STATE BUSINESS SYSTEMS LLC Z-Select 4000T - 2.25" x 1.25" Thermal transfer paper label
Catalog No. NC1825656
Encompass
The Zebra Z-Select 4000T is a thermal transfer, bright white, coated, ultra-smooth, paper label with a permanent acrylic adhesive. Offers excellent adhesion on corrugated packaging. Specially coated for use with compatible Zebra Ribbons for superior durability and image quality. Recommended for a wide variety of applications, such as shipping, receiving, and work-in-process labels.
Catalog No. NC1825656 Supplier TRI STATE BUSINESS SYSTEMS LLC Supplier No. 800272125CS
The Zebra Z-Select 4000T is a thermal transfer, bright white, coated, ultra-smooth, paper label with a permanent acrylic adhesive. Offers excellent adhesion on corrugated packaging. Specially coated for use with compatible Zebra Ribbons for superior durability and image quality. Recommended for a wide variety of applications, such as shipping, receiving, and work-in-process labels.