Learn More
Business Communication Solutions 2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label, Ultra Removable Adhesive, 610 Labels/Roll, 12 Rolls/Case, 0.75" Core, 2.8715" OD.
Supplier:  Business Communication Solutions L43230
Encompass_Preferred
2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label. Backed with an ultra-removable adhesive, for easy repositioning. Offers cleanroom packaging and is ideal for keeping products safe from excess environmental contamination prior to use. Cleaned with an in-line sub-micron, MicroClean 10, web cleaning system, which breaks down the static layer, allowing both sides of label stock to be cleaned ultrasonically. Packaged in a fully-ionized cleanroom in MDPE certified cleanroom bags, which are nitrogen purged, backfilled, and then vacuum sealed in double bags. This ensures that the labels are sealed in a sub-micron environment to maintain the cleanliness level and freshness of the labels. Compatible with Zebra Model ZQ620 and QLn320 Thermal Barcode Printers. Two year expiration.
Catalog No. NC2200729