Learn More
Cayman Chemical 2,2’-Bipyridine-5,5’-dicarboxylic Acid, Purity: ≥98%, CAS: 1802-30-8, 1 g
SDP
Catalog No. 502988139
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of metal-organic frameworks for water oxidation, organic photocatalysis, and carbon dioxide reduction
Catalog No. 50-298-8139 Supplier Cayman Chemical Supplier No. 294721 G
A heterocyclic building block; has been used in the synthesis of metal-organic frameworks for water oxidation, organic photocatalysis, and carbon dioxide reduction