Learn More
Cayman Chemical 2,2’-Biquinoline-4,4’-dicarboxylic Acid (sodium salt), Purity: ≥98%, CAS: 979-88-4, 1 g
SDP
Catalog No. 502991014
Encompass_Preferred
A colorimetric reagent for the detection of copper; with Cu+, forms a stable 2:1 complex that has an absorbance maximum at 562 nm; commonly used as a reagent in the colorimetric BCA protein quantitation assay
Catalog No. 50-299-1014 Supplier Cayman Chemical Supplier No. 304251 G
A colorimetric reagent for the detection of copper; with Cu+, forms a stable 2:1 complex that has an absorbance maximum at 562 nm; commonly used as a reagent in the colorimetric BCA protein quantitation assay