Learn More
Cayman Chemical 2',2'-Difluoro-2'-deoxyuridine, Purity: ≥95%, CAS: 114248-23-6, 10 mg
SDP
Catalog No. 502952405
Encompass_Preferred
An active metabolite of gemcitabine; formed by deamination of gemcitabine by cytidine deaminase in the liver; cytotoxic to HepG2 and A549 cancer cells (IC50s = 3.13 and 3.92 µM, respectively); enhances radiation-induced cell death of ECV304 and NCI H292 cells from 10-100 µM
Catalog No. 50-295-2405 Supplier Cayman Chemical Supplier No. 1168910 MG
An active metabolite of gemcitabine; formed by deamination of gemcitabine by cytidine deaminase in the liver; cytotoxic to HepG2 and A549 cancer cells (IC50s = 3.13 and 3.92 µM, respectively); enhances radiation-induced cell death of ECV304 and NCI H292 cells from 10-100 µM