Learn More
Alkali Scientific 2.2mL 96-Well Deep Well Plate, V Conical Bottom, Square Well, 50/cs
SDP
Catalog No. 502389023
Encompass_Preferred
High chemical stability. Temperature range: -80°C to 120°C. Autoclavable. Centrifugal strength up to 3000g. Compatible with multi-channel pipette and automatic equipment. Optimized bottom design to minimize residual volume. Clear alphanumeric codes for easy identification. Conform to SBS/ ANSI. Specific 96 well deep plate used in nucleic acid extraction. Stackable, reliable tight sealing. Non-pryogenic, DNase/RNase free.
Catalog No. 50-238-9023 Supplier Alkali Scientific Supplier No. MPD29
High chemical stability. Temperature range: -80°C to 120°C. Autoclavable. Centrifugal strength up to 3000g. Compatible with multi-channel pipette and automatic equipment. Optimized bottom design to minimize residual volume. Clear alphanumeric codes for easy identification. Conform to SBS/ ANSI. Specific 96 well deep plate used in nucleic acid extraction. Stackable, reliable tight sealing. Non-pryogenic, DNase/RNase free.