Learn More
Medchemexpress LLC 2-((2S,6S)-2-phenyl-6-hydroxyadamantan-2-yl)-1-(3'-hydroxyazetidin-1-yl)ethanone | 1009583-20-3 | 99.9% | 341.4 g·mol^-1 | C21H27NO3 | 1 ML
SDP
Catalog No. 5000248499
Encompass_Preferred
BMS-816336 is a potent, orally bioavailable inhibitor of human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) with a reported IC50 of ≈3.0 nM. It is supplied for research use in solution (10 mM in DMSO, 1 mL) and in solid forms to support enzymatic and cellular assay development.
  • Potent 11β-HSD1 inhibition with low nanomolar IC50.
  • Suitable for enzymatic and cellular assays.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • Provided in solid quantities for formulation or dose-response studies.
  • High purity suitable for research applications.
Catalog No. 50-002-48499 Supplier Medchemexpress LLC Supplier No. HY10193010MM 1ML
BMS-816336 is a potent, orally bioavailable inhibitor of human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) with a reported IC50 of ≈3.0 nM. It is supplied for research use in solution (10 mM in DMSO, 1 mL) and in solid forms to support enzymatic and cellular assay development.
  • Potent 11β-HSD1 inhibition with low nanomolar IC50.
  • Suitable for enzymatic and cellular assays.
  • Available as a 10 mM solution in DMSO for ready-to-use assays.
  • Provided in solid quantities for formulation or dose-response studies.
  • High purity suitable for research applications.