Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-((2S,6S)-2-phenyl-6-hydroxyadamantan-2-yl)-1-(3'-hydroxyazetidin-1-yl)ethanone | 1009583-20-3 | 99.9% | 341.44 g/mol | C21H27NO3 | 10 MG
SDP

Catalog No. 5000247422
Encompass_Preferred

BMS-816336 is a potent, orally bioavailable small-molecule inhibitor of human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) with an IC50 of 3.0 nM. It is provided for research use and is characterized by a defined molecular formula and molecular weight, with storage recommendations for powder and solutions.

  • Potent inhibitor of human 11β-HSD1 with IC50 3.0 nM.
  • Orally bioavailable small molecule suitable for pharmacology studies.
  • Molecular formula C21H27NO3 and molecular weight 341.44 g/mol.
  • High purity (99.9%) for reliable experimental results.
  • Recommended storage: powder -20°C (long term) and solvent -80°C for stability.

Catalog No. 50-002-47422 Supplier Medchemexpress LLC Supplier No. HY10193010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-816336 is a potent, orally bioavailable small-molecule inhibitor of human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) with an IC50 of 3.0 nM. It is provided for research use and is characterized by a defined molecular formula and molecular weight, with storage recommendations for powder and solutions.

  • Potent inhibitor of human 11β-HSD1 with IC50 3.0 nM.
  • Orally bioavailable small molecule suitable for pharmacology studies.
  • Molecular formula C21H27NO3 and molecular weight 341.44 g/mol.
  • High purity (99.9%) for reliable experimental results.
  • Recommended storage: powder -20°C (long term) and solvent -80°C for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.