Learn More
Medchemexpress LLC 2,3(1H)-Isoquinolinedicarboxylic acid, 3,4-dihydro-, 2-(9H-fluoren-9-ylmethyl) ester, (3R)- | 130309-33-0 | 99.93% | 399.44 | 10 G
SDP
Catalog No. 5000454428
Encompass_Preferred
Fmoc-D-Tic-OH is an amino acid derivative with an Fmoc protecting group. It can be used to synthesize bioactive peptide mimetics, such as [desArg10]HOE 140, which exhibits bradykinin B1 antagonist activity. This product is for research use only and is not sold to patients.
  • Used to synthesize bioactive peptide mimetics
  • Exhibits bradykinin B1 antagonist activity
  • For research use only
Catalog No. 50-004-54428 Supplier Medchemexpress LLC Supplier No. HYW01315310G
Fmoc-D-Tic-OH is an amino acid derivative with an Fmoc protecting group. It can be used to synthesize bioactive peptide mimetics, such as [desArg10]HOE 140, which exhibits bradykinin B1 antagonist activity. This product is for research use only and is not sold to patients.
  • Used to synthesize bioactive peptide mimetics
  • Exhibits bradykinin B1 antagonist activity
  • For research use only