Learn More
Medchemexpress LLC 2-[[3-(2-quinolinylmethoxy)phenyl]amino]benzoic acid hydrochloride | 146662-42-2 | 406.86 g/mol | C23H19ClN2O3 | 5 MG
SDP
Catalog No. 5000240384
Encompass_Preferred
SR2640 hydrochloride is the hydrochloride salt of SR2640, a potent and selective competitive antagonist of leukotriene D4 (LTD4) and leukotriene E4 (LTE4) receptors. It is supplied as a research-grade white to off-white powder for use in in vitro and in vivo studies of leukotriene-mediated signaling, inflammation, and asthma models.
  • Potent and selective leukotriene D4 and E4 receptor antagonist.
  • Suitable for in vitro and in vivo pharmacology and signaling studies.
  • Supplied as a solid powder facilitating compound handling and formulation.
  • Characterized by CAS 146662-42-2 and molecular weight 406.86 g/mol.
  • Useful for investigating leukotriene roles in airway inflammation and asthma models.
  • Provided in small research quantities for dose-ranging experiments.
Catalog No. 50-002-40384 Supplier Medchemexpress LLC Supplier No. HY1076105MG
SR2640 hydrochloride is the hydrochloride salt of SR2640, a potent and selective competitive antagonist of leukotriene D4 (LTD4) and leukotriene E4 (LTE4) receptors. It is supplied as a research-grade white to off-white powder for use in in vitro and in vivo studies of leukotriene-mediated signaling, inflammation, and asthma models.
  • Potent and selective leukotriene D4 and E4 receptor antagonist.
  • Suitable for in vitro and in vivo pharmacology and signaling studies.
  • Supplied as a solid powder facilitating compound handling and formulation.
  • Characterized by CAS 146662-42-2 and molecular weight 406.86 g/mol.
  • Useful for investigating leukotriene roles in airway inflammation and asthma models.
  • Provided in small research quantities for dose-ranging experiments.