Learn More
Echelon Biosciences Research Labs 2,3,22,23-DIOXIDOSQUALENE 1 mg
SDP
Catalog No. 50720760
Encompass
2,3,22,23-Dioxidosqualene (mixture of diastereomers) is a substrate for oxidosqualene cyclase forming 24,25-oxidolanosterol. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-760 Supplier Echelon Biosciences Research Labs Supplier No. S03021MG
2,3,22,23-Dioxidosqualene (mixture of diastereomers) is a substrate for oxidosqualene cyclase forming 24,25-oxidolanosterol. This item is non returnable