Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,3,5-Tetramethylindolenine, 94%

Catalog No. AAH3310909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3310909 10 g
AAH3310918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3310909 Supplier Thermo Scientific Chemicals Supplier No. H3310909
Add to Cart

CAS: 25981-82-2 | C12H15N | 173.26 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25981-82-2
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD03093020
InChI Key RQVAPBRSUHSDGP-UHFFFAOYSA-N
Synonym 2,3,3,5-tetramethyl-3h-indole, 2,3,3,5-tetramethylindolenine, 3h-indole, 2,3,3,5-tetramethyl, pubchem2092, 2,3,3,5-tetramethyl-3h-indol, 2,3,3,5-tetramethyl-3-h-indole, 3h-indole,2,3,3,5-tetramethyl, 2,3,3,5-tetramethyl-3-hydroindole, 2,3,3,5-tetramethyl-3-h indole
PubChem CID 117693
IUPAC Name 2,3,3,5-tetramethylindole
SMILES CC1=NC2=CC=C(C)C=C2C1(C)C

Specifications

Quantity 10 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 173.26
Percent Purity 96%
Chemical Name or Material 2,3,3,5-Tetramethylindolenine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.