Learn More
AdipoGen 2 3 3-Trimethylindolenine
SDP
Supplier:  AdipoGen CDXT0080G050
Encompass_Preferred
Chemical. CAS 1640-39-7. Formula C11H13N. MW 159.23. Synthetic Indole derivative used in the preparation of cyanine dye labelleing reagents and other imaging agents.
Catalog No. 50-256-0025