Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[3-[4-(4-fluorophenyl)-1-piperazinyl]propyl]-2H-naphthyl[1,8-cd]isothiazole-1,1-dioxide | 127625-29-0 | 100.0% | 425.52 g/mol | C23H24FN3O2S | 50 MG
SDP

Catalog No. 5000245940
Encompass_Preferred

Fananserin is a selective 5-hydroxytryptamine2 (5-HT2) receptor antagonist with activity at dopamine D4 receptors, supplied as a 50 mg powder for research use. It is used in pharmacological and neurological studies to probe 5-HT2A and D4 receptor function.

  • High reported purity (≈99.97%).
  • Molecular formula C23H24FN3O2S; molecular weight 425.52 g/mol.
  • Soluble in DMSO: 100 mg/mL (ultrasonic).
  • Powder form with recommended storage -20°C (3 years) or 4°C (2 years).
  • Intended for research use only (RUO); not for human use.

Catalog No. 50-002-45940 Supplier Medchemexpress LLC Supplier No. HY10310450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fananserin is a selective 5-hydroxytryptamine2 (5-HT2) receptor antagonist with activity at dopamine D4 receptors, supplied as a 50 mg powder for research use. It is used in pharmacological and neurological studies to probe 5-HT2A and D4 receptor function.

  • High reported purity (≈99.97%).
  • Molecular formula C23H24FN3O2S; molecular weight 425.52 g/mol.
  • Soluble in DMSO: 100 mg/mL (ultrasonic).
  • Powder form with recommended storage -20°C (3 years) or 4°C (2 years).
  • Intended for research use only (RUO); not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.