Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,4'-Tetrahydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. T159425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T159425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T159425G Supplier TCI America Supplier No. T159425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31127-54-5
Molecular Formula C13H10O5
Molecular Weight (g/mol) 246.218
MDL Number MFCD00083097
InChI Key ZRDYULMDEGRWRC-UHFFFAOYSA-N
PubChem CID 919792
IUPAC Name (4-hydroxyphenyl)-(2,3,4-trihydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=C(C(=C(C=C2)O)O)O)O

Specifications

Melting Point 221°C
Quantity 25 g
Formula Weight 246.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,4′-Tetrahydroxybenzophenone
EINECSNumber (4)-1531
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.