Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4',5',6'-Pentafluoroacetophenone, 97%

Catalog No. AAL0206303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0206303 1 g
AAL0206306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0206303 Supplier Thermo Scientific Chemicals Supplier No. L0206303
Add to Cart

CAS: 652-29-9 | C8H3F5O | 210.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-29-9
Molecular Formula C8H3F5O
Molecular Weight (g/mol) 210.10
MDL Number MFCD00000296
InChI Key FBGHCYZBCMDEOX-UHFFFAOYSA-N
Synonym 2',3',4',5',6'-pentafluoroacetophenone, 1-perfluorophenyl ethanone, pentafluoroacetophenone, 1-pentafluorophenyl ethan-1-one, ethanone, 1-pentafluorophenyl, 2,3,4,5,6-pentafluoroacetophenone, 1-pentafluorphenyl ethanon, 1-acetyl-2,3,4,5,6-pentafluorobenzene, acmc-209npz, 1-pentafluorophenylethanone
PubChem CID 69546
SMILES CC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.476
Boiling Point 130°C to 131°C
Flash Point 65°C (149°F)
Refractive Index 1.4366
Quantity 1 g
Beilstein 2053225
IUPAC Name 1-(2,3,4,5,6-pentafluorophenyl)ethanone
Formula Weight 210.1
Percent Purity 97%
Chemical Name or Material 2',3',4',5',6'-Pentafluoroacetophenone
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 211-487-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.