Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,4,5,6-Pentafluorobenzaldehyde

Catalog No. 501853883
Encompass
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-3883 2.5 g
10-374-810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-3883 Supplier Sigma Aldrich Supplier No. 1037482.5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,3,4,5,6-Pentafluorobenzaldehyde
TRUSTED_SUSTAINABILITY

Specifications

CAS 653-37-2
Melting Point 24°C to 28°C (lit.)
Density 1.588 g/mL (at 25°C (literature))
Boiling Point 164°C to 166°C (lit.)
Molecular Formula C7HF5O
Refractive Index n20/D 1.45 (literature)
Linear Formula C6F5CHO
MDL Number MFCD00003303
Quantity 2.5 g
Molecular Weight (g/mol) 196.07
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.