Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorobenzenesulfonyl chloride, 98+%

Catalog No. AAA1767606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1767606 5 g
AAA1767603 1 g
AAA1767614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1767606 Supplier Thermo Scientific Chemicals Supplier No. A1767606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 832-53-1 | C6ClF5O2S | 266.566 g/mol

Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

Solubility
Soluble in water (reacts slowly).

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases, water/moisture. Keep the container tightly closed in a cool, dry place. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 832-53-1
Molecular Formula C6ClF5O2S
Molecular Weight (g/mol) 266.566
MDL Number MFCD00007427
InChI Key UOJCTEGNHXRPKO-UHFFFAOYSA-N
Synonym pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
PubChem CID 70026
IUPAC Name 2,3,4,5,6-pentafluorobenzenesulfonyl chloride
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F

Specifications

Density 1.796
Boiling Point 210°C to 211°C
Flash Point 90°C (194°F)
Refractive Index 1.479
Quantity 5 g
UN Number UN3265
Beilstein 2142325
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts slowly).
Formula Weight 266.57
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzenesulfonyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-620-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.