Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3,4,5,6-Pentafluorobenzoyl chloride

Catalog No. 1037725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-377-25G 5 g
10-377-21G 1 g
10-377-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-377-25G Supplier Sigma Aldrich Supplier No. 1037725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3,4,5,6-pentafluorobenzoyl chloride was used for derivatization of 5-androstenediol, testosterone, estradiol , dihydrotestosterone and estrone in a study to determine their serum levels
  • 2,3,4,5,6-pentafluorobenzoyl chloride was used to derivatize dimethylamine(dma) in study to develop a rapid and accurate GC-ms method for determination of dma in human urine
  • 2,3,4,5,6-pentafluorobenzoyl chloride was used in a study to detect the presence of enterobacter cloacae in cultures of leuconostoc mesenteroide by gas chromatography/mass spectrometry

Specifications

CAS 2251-50-5
Density 1.601 g/mL (at 25°C (literature))
Boiling Point 158°C to 159°C (lit.)
Molecular Formula C7ClF5O
Refractive Index n20/D 1.453 (literature)
Linear Formula C6F5COCl
MDL Number MFCD00000657
Quantity 5 g
Synonym Pentafluorobenzoic acid chloride; Perfluorobenzoyl chloride
Molecular Weight (g/mol) 230.52
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.