Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorostyrene (stabilized with TBC) 98.0+%, TCI America™
SDP

Catalog No. P086225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P086225G 25 g
P08625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P086225G Supplier TCI America Supplier No. P086225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 653-34-9
Molecular Formula C8H3F5
Molecular Weight (g/mol) 194.104
MDL Number MFCD00000300
InChI Key LVJZCPNIJXVIAT-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorostyrene, pentafluorostyrene, benzene, ethenylpentafluoro, 1,2,3,4,5-pentafluoro-6-vinyl-benzene, 2',3',4',5',6'-pentafluorostyrene, 2-2,3,4,5,6-pentafluorophenyl ethyl, vinylpentafluorobenzene, acmc-1avmt, styrene, 2,3,4,5,6-pentafluoro, 1,2,3,4,5-pentafluoro-6-vinylbenzene
PubChem CID 69556
IUPAC Name 1-ethenyl-2,3,4,5,6-pentafluorobenzene
SMILES C=CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
UN Number 1993
Formula Weight 194.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5,6-Pentafluorostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.