Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorotoluene, 99%

Catalog No. AAB2141918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2141918 50 g
AAB2141909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2141918 Supplier Thermo Scientific Chemicals Supplier No. B2141918
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 771-56-2 | C7H3F5 | 182.093 g/mol

2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.

Solubility
It is soluble in most common organic solvents and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-56-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.093
MDL Number MFCD00000298
InChI Key SXPRVMIZFRCAGC-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene
PubChem CID 69869
IUPAC Name 1,2,3,4,5-pentafluoro-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point -30°C
Density 1.439
Boiling Point 116°C to 117°C
Flash Point 34°C (93°F)
Refractive Index 1.403
Quantity 50 g
UN Number UN1993
Beilstein 1911885
Solubility Information It is soluble in most common organic solvents and insoluble in water.
Formula Weight 182.09
Percent Purity 99%
Chemical Name or Material 2,3,4,5,6-Pentafluorotoluene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-233-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.