Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4',5',6'-Pentamethylacetophenone, 98+%, Thermo Scientific™

Catalog No. AAA1904014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1904014 25 g
AAA1904006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1904014 Supplier Thermo Scientific Chemicals Supplier No. A1904014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-01-9
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00014986
InChI Key CTTYWXDVWGKHKJ-UHFFFAOYSA-N

Specifications

Melting Point 84°C to 87°C
Quantity 25 g
Beilstein 1867535
Synonym 2',3',4',5',6'-pentamethylacetophenone, 1-pentamethylphenyl ethanone, 1-pentamethylphenyl ethan-1-one, 2,3,4,5,6-pentamethylacetophenone, 1-2,3,4,5,6-pentamethylphenyl ethanone, acmc-1cl24, 2,3,4,5,6-pentamethyl acetophenone, 1,2,3,4,5-pentamethyl-6-acetylbenzene, 1-acetyl-2,3,4,5,6-pentamethylbenzene, ethanone,1-2,3,4,5,6-pentamethylphenyl
SMILES CC1=C(C(=C(C(=C1C)C)C(=O)C)C)C
IUPAC Name 1-(2,3,4,5,6-pentamethylphenyl)ethanone
PubChem CID 74876
Formula Weight 190.29
Percent Purity ≥98%
Chemical Name or Material 2',3',4',5',6'-Pentamethylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.