Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3,4,5-Tetracaffeoyl-D-Glucaric acid | 1419478-52-6 | 5 MG
SDP

Catalog No. 5000419395
Encompass_Preferred

2,3,4,5-Tetracaffeoyl-D-Glucaric acid is a caffeoyl-D-glucaric acid derivative isolated from the Genus Gnaphalium, intended for research use only.

  • Isolated from the genus Gnaphalium
  • Purity of 99.29%
  • Molecular weight of 858.71
  • Formula: C42H34O20
  • Appears as an off-white to light yellow solid
  • Initial source includes various plants
  • Classified as phenylpropanoid
  • Soluble in DMSO at 100 mg/mL
  • Used in inflammation/immunology research

Catalog No. 50-004-19395 Supplier Medchemexpress LLC Supplier No. HYN43115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3,4,5-Tetracaffeoyl-D-Glucaric acid is a caffeoyl-D-glucaric acid derivative isolated from the Genus Gnaphalium, intended for research use only.

  • Isolated from the genus Gnaphalium
  • Purity of 99.29%
  • Molecular weight of 858.71
  • Formula: C42H34O20
  • Appears as an off-white to light yellow solid
  • Initial source includes various plants
  • Classified as phenylpropanoid
  • Soluble in DMSO at 100 mg/mL
  • Used in inflammation/immunology research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.