Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrachlorobenzoic Acid 97.0+%, TCI America™
SDP

Catalog No. T237825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T237825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T237825G Supplier TCI America Supplier No. T237825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-74-8
Molecular Formula C7H2Cl4O2
Molecular Weight (g/mol) 259.89
MDL Number MFCD00061126
InChI Key FUXMTEKFGUWSNC-UHFFFAOYSA-N
PubChem CID 5782
IUPAC Name 2,3,4,5-tetrachlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 195°C
Color Red-White
Quantity 25 g
Formula Weight 259.89
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5-Tetrachlorobenzoic Acid
EINECSNumber (3)-0062
RTECSNumber DH6833000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.