Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrachlorophenol, MilliporeSigma™ Supelco™

Catalog No. 111018881 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8881 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8881 Supplier MilliporeSigma Supelco Supplier No. 442281
Add to Cart
Add to Cart

2,3,4,5-Tetrachlorophenol belongs to the class of chlorophenol (CP) compounds, regarded as persistent and potentially hazardous substances. It finds its use as a fungicide, and in applications such as preservation of plants, and sewage treatment.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 4901-51-3
Physical Form Neat
Linear Formula C6H2 Cl4 O
MDL Number MFCD00053449
Type Organic Pollutant Standard
Quantity 100 mg
Formula Weight 231.89
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.