Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3,4,5-Tetrafluoro-4″-propyl-1,1':4',1″-terphenyl 98.0+%, TCI America™
SDP

Catalog No. T33185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33185G 5 g
T33181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33185G Supplier TCI America Supplier No. T33185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 205806-87-7
Molecular Formula C21H16F4
Molecular Weight (g/mol) 344.35
MDL Number MFCD12964272
InChI Key YBPXWWAJMUUUAV-UHFFFAOYSA-N
PubChem CID 12010664
IUPAC Name 2,3',4',5'-tetrafluoro-4-(4-propylphenyl)-1,1'-biphenyl
SMILES CCCC1=CC=C(C=C1)C1=CC(F)=C(C=C1)C1=CC(F)=C(F)C(F)=C1

Specifications

Color White
Quantity 5 g
Synonym 2′,3,4,5-Tetrafluoro-4′′-propyl-p-terphenyl
Formula Weight 344.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3,4,5-Tetrafluoro-4′′-propyl-1,1′:4′,1′′-terphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.