Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluoro-6-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T186225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T186225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T186225G Supplier TCI America Supplier No. T186225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16583-08-7
Molecular Formula C7HF4NO4
Molecular Weight (g/mol) 239.082
MDL Number MFCD02093916
InChI Key JQGBMYKEWLSLCI-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro-6-nitrobenzoic acid, 6-nitro-2,3,4,5-tetrafluorobenzoic acid, benzoicacid, 2,3,4,5-tetrafluoro-6-nitro, benzoic acid, 2,3,4,5-tetrafluoro-6-nitro, acmc-1c6gj, intermediates-zcf02245, 2,3,4,5-tetrafluoro-6-nitrobenzoicacid, 6-nitro-2,3,4,5-tetrafluorobenzoicacid, 2-nitro-3,4,5,6-tetrafluorobenzoic acid, 3,4,5,6-tetrafluoro-2-nitrobenzoic acid
PubChem CID 2733983
SMILES C1(=C(C(=C(C(=C1F)F)F)F)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 139°C
Color Yellow
Quantity 25 g
Formula Weight 239.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5-Tetrafluoro-6-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.