Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T13955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13955G 5 g
T139525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13955G Supplier TCI America Supplier No. T13955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1201-31-6
Molecular Formula C7H2F4O2
Molecular Weight (g/mol) 194.09
MDL Number MFCD00009613
InChI Key SFKRXQKJTIYUAG-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluoro benzoic acid, benzoic acid, tetrafluoro, pubchem1350, tetrafluorobenzoic acid, acmc-1bre4, ksc174o7d, sfkrxqkjtiyuag-uhfffaoysa, 2,3,4,5-tetrafluorbenzoic acid, 2,3,4,5-tetrafluoro-benzoic acid, 2,3,4,5-?tetrafluorobenzoic acid
PubChem CID 297549
IUPAC Name 2,3,4,5-tetrafluorobenzoic acid
SMILES OC(=O)C1=CC(F)=C(F)C(F)=C1F

Specifications

Melting Point 87°C
Quantity 5 g
Formula Weight 194.08
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,5-Tetrafluorobenzoic Acid
EINECSNumber (3)-3967
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.