Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5-Tetrafluorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. T18535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18535G Supplier TCI America Supplier No. T18535G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 654-53-5
Molecular Formula C7HF7
Molecular Weight (g/mol) 218.074
MDL Number MFCD01862013
InChI Key ZWVOHERGWLDGFT-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl benzene, benzene, tetrafluoro trifluoromethyl, alpha,alpha,alpha,2,3,4,5-heptafluorotoluene, acmc-209trj, 3,4,5,6-tetrafluorobenzotrifluoride, 2,3,4,5-tetrafluoro-benzotrifluoride, 1,2,3,4-tetrafluoro-5-trifluoromethyl-benzene, 2,3,4,5-tetrafluoro-1-trifluoromethyl benzene
PubChem CID 2733690
IUPAC Name 1,2,3,4-tetrafluoro-5-(trifluoromethyl)benzene
SMILES C1=C(C(=C(C(=C1F)F)F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
UN Number 1993
Formula Weight 218.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,5-Tetrafluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.