Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine | 69718-72-5 | 99.8% | 159.23 | 1 G
SDP

Catalog No. 5000405080
Encompass_Preferred

2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate for the synthesis of various active compounds.

  • Purity of 99.78%
  • Molecular formula C11H13N
  • Appearance as a solid-liquid mixture
  • Light yellow to brown color
  • Store at 4°C, protect from light; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Classified as a drug intermediate
  • For research use only

Catalog No. 50-004-05080 Supplier Medchemexpress LLC Supplier No. HY200071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate for the synthesis of various active compounds.

  • Purity of 99.78%
  • Molecular formula C11H13N
  • Appearance as a solid-liquid mixture
  • Light yellow to brown color
  • Store at 4°C, protect from light; in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Classified as a drug intermediate
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.