Learn More
Medchemexpress LLC 2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine | 69718-72-5 | 99.8% | 159.23 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY20007
Encompass_Preferred
2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate used for the synthesis of various active compounds. This product is for research use only.
  • Used for synthesis of various active compounds
  • For research use only
  • Store at 4°C, protected from light
  • Documentation includes Data Sheet, COA, and SDS
Catalog No. 50-004-02908