Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl 2,2,2-Trichloroacetimidate 95.0+%, TCI America™
SDP

Catalog No. T22951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22951G 1 g
T22955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22951G Supplier TCI America Supplier No. T22951G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 86520-63-0 | C16H20Cl3NO10 | 492.683 g/mol

Chemical Identifiers

CAS 86520-63-0
Molecular Formula C16H20Cl3NO10
Molecular Weight (g/mol) 492.683
MDL Number MFCD07369652
InChI Key IBUZGVQIKARDAF-MBJXGIAVSA-N
PubChem CID 11092215
IUPAC Name [(2R,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-(2,2,2-trichloroethanimidoyl)oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=N)C(Cl)(Cl)Cl)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 122°C
Color Yellow
Quantity 1 g
Formula Weight 492.68
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl 2,2,2-Trichloroacetimidate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.