Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl Bromide (stabilized with CaCO3) 98.0+%, TCI America™
SDP

Catalog No. T19615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19615G Supplier TCI America Supplier No. T19615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 572-09-8
Molecular Formula C14H19BrO9
Molecular Weight (g/mol) 411.20
MDL Number MFCD00063254
InChI Key CYAYKKUWALRRPA-HAGGZJDPNA-N
Synonym 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, acetobromglucose, acetobromoglucose, acetobromo-alpha-d-glucose, a-acetobromglucose, acetobromo glucose, 2,3,4,6-tetra-o-acetyl-, a-d-glucopyranosyl bromide, 2r,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxan-2-yl methyl acetate, 2r,3r,4s,5r,6r-3,4,5-triacetyloxy-6-bromooxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-alpha-d-glucopyranosyl bromide, stab. with calcium carbonate
PubChem CID 101776
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Formula Weight 411.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-alpha-D-glucopyranosyl Bromide (stabilized with CaCO3)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.