Learn More
Medchemexpress LLC 2,3,4,6-tetra-O-acetyl-α-D-galactopyranosyl bromide | 3068-32-4 | MFCD00063686 | 97.0% | 411.20 g·mol⁻¹ | C14H19BrO9 | 5 G
SDP
Catalog No. 5000301717
Encompass_Preferred
Acetobromo-α-D-galactose is a brominating reagent used in carbohydrate chemistry as an intermediate for the synthesis of glycosides and oligosaccharides. It is supplied for research use and is supported by analytical documentation including COA, data sheet, SDS, and 1H NMR.
Catalog No. 50-003-01717 Supplier Medchemexpress LLC Supplier No. HYW0288135G
Acetobromo-α-D-galactose is a brominating reagent used in carbohydrate chemistry as an intermediate for the synthesis of glycosides and oligosaccharides. It is supplied for research use and is supported by analytical documentation including COA, data sheet, SDS, and 1H NMR.