Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013074 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3074 100 mg
11-101-3073 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3074 Supplier MilliporeSigma Supelco Supplier No. T5783100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate is a chiral derivatization reagent which reacts mainly with enantiomeric amino acids.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00043085
Synonym GITC

Specifications

Melting Point 114°C to 116°C
CAS 14152-97-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C15H19 NO9 S
Quantity 100 mg
Molecular Weight (g/mol) 389.38
Formula Weight 389.38
Percent Purity ≥98.0% (HPLC)
Grade For chiral derivatization
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.