Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzoyl-alpha-D-glucopyranosyl p-Trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate 96.0+%, TCI America™
SDP

Catalog No. T1991200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1991200MG 200 mg
T19911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1991200MG Supplier TCI America Supplier No. T1991200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 428816-48-2 | C50H37F6NO10S | 957.893 g/mol

Chemical Identifiers

CAS 428816-48-2
Molecular Formula C50H37F6NO10S
Molecular Weight (g/mol) 957.893
MDL Number MFCD06797174
InChI Key HTRJVXKRCWWVSR-KPAAGLOJSA-N
PubChem CID 11378098
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-[N-[4-(trifluoromethyl)phenyl]-C-[[4-(trifluoromethyl)phenyl]methylsulfanyl]carbonimidoyl]oxyoxan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)OC(=NC3=CC=C(C=C3)C(F)(F)F)SCC4=CC=C(C=C4)C(F)(F)F)OC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)OC(=O)C7=CC=CC=C7

Specifications

Color White
Quantity 200 mg
Formula Weight 957.89
Percent Purity ≥96.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzoyl-alpha-D-glucopyranosyl p-Trifluoromethylbenzylthio-N-(p-trifluoromethylpheny
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.