Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzyl-D-glucopyranosyl Fluoride 96.0+%, TCI America™
SDP

Catalog No. T1971500MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1971500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1971500MG Supplier TCI America Supplier No. T1971500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122741-44-0
Molecular Formula C34H35FO5
Molecular Weight (g/mol) 542.65
MDL Number MFCD01862264
InChI Key QNXIKNZDQVSBCO-UHFFFAOYNA-N
PubChem CID 133556312
IUPAC Name 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-fluorooxane
SMILES FC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 500 mg
Formula Weight 542.65
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-glucopyranosyl Fluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.