Learn More
Medchemexpress LLC 2-(3,4-dihydroquinolin-1(2H)-yl)-N-(5-methyl-1,2-oxazol-3-yl)-2-phenylacetamide | 1031496-06-6 | MFCD00045972 | 99.9% | 347.41 g/mol | C21H21N3O2 | 5 MG
SDP
Catalog No. 5000264305
Encompass_Preferred
CIM0216 is a potent, selective agonist of the TRPM3 ion channel used as a research tool to probe TRPM3-dependent cellular and neurophysiological responses, including neurogenic inflammation and sensory neuron signaling. It is supplied as a high-purity solid or as a 10 mM solution in DMSO for laboratory use.
  • Potent TRPM3 agonist with selectivity over related TRP channels.
  • Induces TRPM3-dependent pain responses and neuropeptide release in vitro.
  • High purity (99.94%) suitable for biochemical and cellular assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 347.41 g/mol; formula C21H21N3O2.
Catalog No. 50-002-64305 Supplier Medchemexpress LLC Supplier No. HY1102205MG
CIM0216 is a potent, selective agonist of the TRPM3 ion channel used as a research tool to probe TRPM3-dependent cellular and neurophysiological responses, including neurogenic inflammation and sensory neuron signaling. It is supplied as a high-purity solid or as a 10 mM solution in DMSO for laboratory use.
  • Potent TRPM3 agonist with selectivity over related TRP channels.
  • Induces TRPM3-dependent pain responses and neuropeptide release in vitro.
  • High purity (99.94%) suitable for biochemical and cellular assays.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Molecular weight 347.41 g/mol; formula C21H21N3O2.