Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(3,4-dihydroxy-5-nitrophenyl)methylidene]propanedinitrile | 116313-73-6 | 98.7% | 231.16 g/mol | C10H5N3O4 | 10 MG
SDP

Catalog No. 5000247968
Encompass_Preferred

Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in research to block signaling pathways and reduce inflammatory responses, notably by inhibiting ICAM1 expression and TNFα-mediated cytotoxicity in vitro.

  • Potent tyrosine kinase inhibitor.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro.
  • Molecular formula C10H5N3O4; molecular weight 231.16 g/mol.
  • Purity 98.7% as supplied.
  • Soluble ≥125 mg/mL (≈540.75 mM) in DMSO.
  • Appearance solid, light yellow to yellow.
  • Recommended storage: powder -20°C up to 3 years or 4°C up to 2 years; in solution -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-47968 Supplier Medchemexpress LLC Supplier No. HY10196110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in research to block signaling pathways and reduce inflammatory responses, notably by inhibiting ICAM1 expression and TNFα-mediated cytotoxicity in vitro.

  • Potent tyrosine kinase inhibitor.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro.
  • Molecular formula C10H5N3O4; molecular weight 231.16 g/mol.
  • Purity 98.7% as supplied.
  • Soluble ≥125 mg/mL (≈540.75 mM) in DMSO.
  • Appearance solid, light yellow to yellow.
  • Recommended storage: powder -20°C up to 3 years or 4°C up to 2 years; in solution -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.