Learn More
Medchemexpress LLC 2-[(3,4-dihydroxy-5-nitrophenyl)methylidene]propanedinitrile | 116313-73-6 | 98.7% | 231.16 g/mol | C10H5N3O4 | 25 MG
SDP
Catalog No. 5000247989
Encompass_Preferred
Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in biochemical and cellular research to probe tyrosine kinase-dependent signaling pathways, ICAM1 expression, and TNFα-mediated cytotoxicity. It is supplied as a high-purity research reagent intended for in vitro assays.
  • High purity suitable for analytical and cell-based assays.
  • Useful for investigating tyrosine kinase signaling pathways.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro.
  • Supplied as a stable, solid reagent for easy handling and storage.
  • Batch-tested with a certificate of analysis for lot verification.
  • Compatible with common solvents for assay formulation.
Catalog No. 50-002-47989 Supplier Medchemexpress LLC Supplier No. HY10196125MG
Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used in biochemical and cellular research to probe tyrosine kinase-dependent signaling pathways, ICAM1 expression, and TNFα-mediated cytotoxicity. It is supplied as a high-purity research reagent intended for in vitro assays.
  • High purity suitable for analytical and cell-based assays.
  • Useful for investigating tyrosine kinase signaling pathways.
  • Inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro.
  • Supplied as a stable, solid reagent for easy handling and storage.
  • Batch-tested with a certificate of analysis for lot verification.
  • Compatible with common solvents for assay formulation.