Learn More
Medchemexpress LLC 2-(3,4-dimethoxyphenyl)-5-fluoro-1,3-benzothiazole | 872726-44-8 | MFCD12024619 | 99.8% | 289.32 g/mol | C15H12FNO2S | 100 MG
SDP
Catalog No. 5000228495
Encompass_Preferred
GW 610 is a fluorinated 2-arylbenzothiazole antitumor agent and a potent aryl hydrocarbon receptor (AhR) ligand. It induces CYP1A1 expression and demonstrates selective cytotoxic activity against lung, colon, and breast cancer cell lines. This compound is intended for research use in vitro.
Catalog No. 50-002-28495 Supplier Medchemexpress LLC Supplier No. HY118474100MG
GW 610 is a fluorinated 2-arylbenzothiazole antitumor agent and a potent aryl hydrocarbon receptor (AhR) ligand. It induces CYP1A1 expression and demonstrates selective cytotoxic activity against lung, colon, and breast cancer cell lines. This compound is intended for research use in vitro.