Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole | 872726-44-8 | MFCD12024619 | 99.8% | 289.32 g/mol | C15H12FNO2S | 10 MG
SDP

Catalog No. 5000227647
Encompass_Preferred

GW 610 (2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole) is an antitumor benzothiazole small molecule and a potent aryl hydrocarbon receptor (AhR) ligand. Supplied for research use only, the compound is provided as a 10 mg solid suitable for in vitro and preclinical studies.

  • High purity: 99.8% (manufacturer reported).
  • Molecular weight: 289.32 g/mol, useful for dosing calculations.
  • Chemical formula: C15H12FNO2S.
  • Shown in literature to have selective antiproliferative activity against cancer cell lines.
  • Suitable for mechanistic studies involving AhR signaling.

Catalog No. 50-002-27647 Supplier Medchemexpress LLC Supplier No. HY11847410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GW 610 (2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole) is an antitumor benzothiazole small molecule and a potent aryl hydrocarbon receptor (AhR) ligand. Supplied for research use only, the compound is provided as a 10 mg solid suitable for in vitro and preclinical studies.

  • High purity: 99.8% (manufacturer reported).
  • Molecular weight: 289.32 g/mol, useful for dosing calculations.
  • Chemical formula: C15H12FNO2S.
  • Shown in literature to have selective antiproliferative activity against cancer cell lines.
  • Suitable for mechanistic studies involving AhR signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.