Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole | 872726-44-8 | MFCD12024619 | 99.8% | 289.32 g/mol | C15H12FNO2S | 5 MG
SDP

Catalog No. 5000228001
Encompass_Preferred

GW 610 is a fluorinated 2-arylbenzothiazole antitumor agent used in research to study aryl hydrocarbon receptor (AhR)-mediated pathways and anticancer activity in cell lines. It induces CYP1A1 expression in MCF-7 cells and shows selective growth-inhibitory effects against lung, colon, and breast cancer cell lines.

  • Potent aryl hydrocarbon receptor ligand, useful for AhR pathway studies.
  • Induces CYP1A1 expression in MCF-7 cells.
  • Demonstrates selective anticancer activity against lung, colon, and breast cancer cell lines.
  • High chemical purity reported (99.8%), suitable for research use.
  • Molecular weight 289.32 g/mol; chemical formula C15H12FNO2S.
  • Store at 4°C protected from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-28001 Supplier Medchemexpress LLC Supplier No. HY1184745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GW 610 is a fluorinated 2-arylbenzothiazole antitumor agent used in research to study aryl hydrocarbon receptor (AhR)-mediated pathways and anticancer activity in cell lines. It induces CYP1A1 expression in MCF-7 cells and shows selective growth-inhibitory effects against lung, colon, and breast cancer cell lines.

  • Potent aryl hydrocarbon receptor ligand, useful for AhR pathway studies.
  • Induces CYP1A1 expression in MCF-7 cells.
  • Demonstrates selective anticancer activity against lung, colon, and breast cancer cell lines.
  • High chemical purity reported (99.8%), suitable for research use.
  • Molecular weight 289.32 g/mol; chemical formula C15H12FNO2S.
  • Store at 4°C protected from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.