Learn More
Medchemexpress LLC 2-(3,4-dimethoxyphenyl)benzo[h]chromen-4-one | 14756-24-2 | MFCD00143008 | 98.5% | 332.35 | C21H16O4 | 50 MG
SDP
Catalog No. 5000247015
Encompass_Preferred
DiMNF (3',4'-Dimethoxy-αNF) is a small-molecule aryl hydrocarbon receptor (AHR) modulator supplied as a light yellow to yellow solid for research use. It has molecular formula C21H16O4, molecular weight 332.35, and CAS 14756-24-2. Provided in small packaged quantities for in vitro and exploratory studies; not for clinical use.
  • Used as an AHR modulator in biochemical and cell-based assays.
  • High purity suitable for research applications.
  • Available in small package sizes for laboratory studies.
  • Solid, light yellow to yellow appearance for easy handling.
  • Supplied for research use only; not for clinical or human use.
Catalog No. 50-002-47015 Supplier Medchemexpress LLC Supplier No. HY10322250MG
DiMNF (3',4'-Dimethoxy-αNF) is a small-molecule aryl hydrocarbon receptor (AHR) modulator supplied as a light yellow to yellow solid for research use. It has molecular formula C21H16O4, molecular weight 332.35, and CAS 14756-24-2. Provided in small packaged quantities for in vitro and exploratory studies; not for clinical use.
  • Used as an AHR modulator in biochemical and cell-based assays.
  • High purity suitable for research applications.
  • Available in small package sizes for laboratory studies.
  • Solid, light yellow to yellow appearance for easy handling.
  • Supplied for research use only; not for clinical or human use.