Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(3,4-epoxycyclohexyl)ethyltriethoxysilane

Catalog No. CDS0105271G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS0105271G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS0105271G Supplier Sigma Aldrich Supplier No. CDS0105271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methylbenzylamine ({(3-chlorophenyl)methanamine}) has been used as a templating agent in the synthesis of amine-templated materials containing two-dimensional lithium beryllofluoride sheets of the stoichiometry [libef(4)](-)
  • It has also been used to study the chromatographic behavior of a series of substituted aniline and pyridine basic compounds on c18 bonded silica

Specifications

CAS 10217-34-2
Molecular Formula C14H28O4Si
Linear Formula C14H28O4Si
MDL Number MFCD00069159
Quantity 5 g
Molecular Weight (g/mol) 288.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.