Learn More
Medchemexpress LLC 2-((3-(4-ethoxyphenyl)-1,2,4-triazolo[4,3-b]pyridazin-6-yl)thio)-N-(4-fluorophenyl)acetamide | 721964-51-8 | 98.3% | 423.46 | C21H18FN5O2S | 5mg
SDP
Catalog No. 5000270269
Encompass_Preferred
Protein kinase inhibitor 11 (Compound I-96) is a protein kinase inhibitor that can inhibit the activity of PIM-1 CDK-2 GSK-3 and SRC Protein kinase inhibitor 11 holds promise for research in cancer immune disorders and neurodegenerative diseases[1]
Catalog No. 50-002-70269 Supplier Medchemexpress LLC Supplier No. HYW8445585MG
Protein kinase inhibitor 11 (Compound I-96) is a protein kinase inhibitor that can inhibit the activity of PIM-1 CDK-2 GSK-3 and SRC Protein kinase inhibitor 11 holds promise for research in cancer immune disorders and neurodegenerative diseases[1]