Learn More
Medchemexpress LLC 2-(3-(4-fluoro-3-pyridin-3-yl-phenyl)-imidazo(1,2-a)pyrimidin-7-yl)-propan-2-ol | 461449-78-5 | 97.0% | 348.37 g·mol⁻1 | C20H17FN4O | 10MG
SDP
Catalog No. 5000236388
Encompass_Preferred
MRK-623 is an orally active, high-affinity GABAA receptor agonist research compound used to study GABAergic signaling and anxiolytic effects in vitro and in vivo. It is supplied as a white to off-white solid with molecular formula C20H17FN4O and molecular weight 348.37 g·mol⁻1, and is provided in small research pack sizes for biochemical and pharmacological assays.
  • Orally active GABAA receptor agonist for GABAergic research.
  • High affinity against multiple GABAA subunits with low-nanomolar potency.
  • High purity suitable for research (manufacturer COA reported 97.0% by NMR).
  • Available in small research quantities, including 10 mg packs.
  • White to off-white solid physical form, soluble in appropriate solvents for assays.
  • Recommended storage: powder at -20°C or 4°C; store solutions at -80°C for long term.
Catalog No. 50-002-36388 Supplier Medchemexpress LLC Supplier No. HY12384010MG
MRK-623 is an orally active, high-affinity GABAA receptor agonist research compound used to study GABAergic signaling and anxiolytic effects in vitro and in vivo. It is supplied as a white to off-white solid with molecular formula C20H17FN4O and molecular weight 348.37 g·mol⁻1, and is provided in small research pack sizes for biochemical and pharmacological assays.
  • Orally active GABAA receptor agonist for GABAergic research.
  • High affinity against multiple GABAA subunits with low-nanomolar potency.
  • High purity suitable for research (manufacturer COA reported 97.0% by NMR).
  • Available in small research quantities, including 10 mg packs.
  • White to off-white solid physical form, soluble in appropriate solvents for assays.
  • Recommended storage: powder at -20°C or 4°C; store solutions at -80°C for long term.