Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4'-Trichloroacetophenone 95%, Thermo Scientific™

Catalog No. AC176390500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176390500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176390500 Supplier Thermo Scientific Chemicals Supplier No. 176390500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13608-87-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
InChI Key BXJZZJYNVIDEKG-UHFFFAOYSA-N
Synonym 2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
PubChem CID 26132
IUPAC Name 1-(2,3,4-trichlorophenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Linear Formula Cl3C6H2COCH3
Quantity 50 g
Formula Weight 223.49
Chemical Name or Material 2', 3', 4'-Trichloroacetophenone, 95%
Hazard Category LACHRYMATOR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.