Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4'-Trichloroacetophenone 98.0+%, TCI America™
SDP

Catalog No. T23065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23065G 5 g
T230625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23065G Supplier TCI America Supplier No. T23065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13608-87-2
Molecular Formula C8H5Cl3O
Molecular Weight (g/mol) 223.477
MDL Number MFCD00000542
InChI Key BXJZZJYNVIDEKG-UHFFFAOYSA-N
PubChem CID 26132
IUPAC Name 1-(2,3,4-trichlorophenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)Cl)Cl)Cl

Specifications

Melting Point 65°C
Boiling Point 144°C
Quantity 5 g
Synonym 2',3',4'-trichloroacetophenone, 1-2,3,4-trichlorophenyl ethanone, 2,3,4-trichloroacetophenone, ethanone, 1-2,3,4-trichlorophenyl, 1-2,3,4-trichlorophenyl ethan-1-one, ccris 785, unii-8hvd33n785, acetophenone, 2',3',4'-trichloro, ethanone,1-2,3,4-trichlorophenyl, 1-acetyl-2,3,4-trichlorobenzene
Formula Weight 223.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′,4′-Trichloroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.