Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trichloronitrobenzene 99.0+%, TCI America™
SDP

Catalog No. T1197500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1197500G 500 g
T119725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1197500G Supplier TCI America Supplier No. T1197500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17700-09-3
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00007063
InChI Key BGKIECJVXXHLDP-UHFFFAOYSA-N
Synonym 2,3,4-trichloronitrobenzene, trichloronitrobenzene, 2,3,4-trichloro-1-nitrobenzene, 4-nitro-1,2,3-trichlorobenzene, benzene, 1,2,3-trichloro-4-nitro, benzene, 4-nitro-1,2,3-trichloro, 1,2,3-trichloronitrobenzene, 1,2,3-tris chloranyl-4-nitro-benzene, pubchem8437, acmc-1bpdn
PubChem CID 28697
IUPAC Name 1,2,3-trichloro-4-nitrobenzene
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])Cl)Cl)Cl

Specifications

Melting Point 56°C
Quantity 500 g
Formula Weight 226.44
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trichloronitrobenzene
RTECSNumber DA6612000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.